siRNA Details
| Gene Symbol: Fam133b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024799.1 | NP_001019970.1 | 8 | 576 | ||
| XM_008762708.1 | XP_008760930.1 | 8 | 667 | |||
| GenBank mRNA | BC094528.1 | - | 8 | 576 | 7320019 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam133b| Gene Symbol: Fam133b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024799.1 | NP_001019970.1 | 8 | 576 | ||
| XM_008762708.1 | XP_008760930.1 | 8 | 667 | |||
| GenBank mRNA | BC094528.1 | - | 8 | 576 | 7320019 | |