siRNA Details
| Gene Symbol: Fam133b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024799.1 | NP_001019970.1 | 7, 8 | 546 | ||
| XM_008762708.1 | XP_008760930.1 | 7, 8 | 637 | |||
| GenBank mRNA | BC094528.1 | - | 7, 8 | 546 | 7320019 | |

Fam133b
Genomic Map