siRNA Details
| Gene Symbol: Mios | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173554.1 | NP_001167025.1 | 3 | 680 | ||
| XM_006236090.2 | XP_006236152.1 | 2 | 680 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mios| Gene Symbol: Mios | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173554.1 | NP_001167025.1 | 3 | 680 | ||
| XM_006236090.2 | XP_006236152.1 | 2 | 680 | |||