siRNA Details
| Gene Symbol: Mdfic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105668.1 | NP_001099138.1 | 4 | 803 | ||
| XM_006236102.2 | XP_006236164.1 | 4 | 1079 | |||
| XM_006236103.2 | XP_006236165.1 | 4 | 1108 | |||
| GenBank mRNA | CV106654.1 | - | 604 | |||

Mdfic
Genomic Map