siRNA Details
| Gene Symbol: Ccdc136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006236239.2 | XP_006236301.2 | 6, 7 | 1123 | ||
| XM_006236240.2 | XP_006236302.1 | 5, 6 | 887 | |||
| XM_006236241.2 | XP_006236303.2 | 6, 7 | 1123 | |||
| XM_006236242.2 | XP_006236304.2 | 6, 7 | 1123 | |||
| XM_008762835.1 | XP_008761057.1 | 6, 7 | 1123 | |||
| XM_008762836.1 | XP_008761058.1 | 6, 7 | 1096 | |||
| XM_008762837.1 | XP_008761059.1 | 6, 7 | 1123 | |||
| XM_008762838.1 | XP_008761060.1 | 6, 7 | 1123 | |||
| XM_008762839.1 | XP_008761061.1 | 6, 7 | 1123 | |||
| GenBank mRNA | FQ228611.1 | - | 3, 4 | 600 | ||

Ccdc136
Genomic Map