siRNA Details
Gene Symbol: Ccdc136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006236239.2 | XP_006236301.2 | 14 | 2769 | ||
XM_006236240.2 | XP_006236302.1 | 13 | 2533 | |||
XM_006236241.2 | XP_006236303.2 | 12 | 2349 | |||
XM_006236242.2 | XP_006236304.2 | 14 | 2769 | |||
XM_008762835.1 | XP_008761057.1 | 14 | 2769 | |||
XM_008762836.1 | XP_008761058.1 | 14 | 2742 | |||
XM_008762837.1 | XP_008761059.1 | 14 | 2769 | |||
GenBank mRNA | FQ228611.1 | - | 11 | 2246 |
