siRNA Details
| Gene Symbol: Ccdc136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006236239.2 | XP_006236301.2 | 2 | 274 | ||
| XM_006236240.2 | XP_006236302.1 | 1 | 38 | |||
| XM_006236241.2 | XP_006236303.2 | 2 | 274 | |||
| XM_006236242.2 | XP_006236304.2 | 2 | 274 | |||
| XM_008762835.1 | XP_008761057.1 | 2 | 274 | |||
| XM_008762836.1 | XP_008761058.1 | 2 | 247 | |||
| XM_008762837.1 | XP_008761059.1 | 2 | 274 | |||
| XM_008762838.1 | XP_008761060.1 | 2 | 274 | |||
| XM_008762839.1 | XP_008761061.1 | 2 | 274 | |||

Ccdc136
Genomic Map