siRNA Details
Gene Symbol: Zc3hav1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_342661.6 | XP_342662.5 | 3 | 1590 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zc3hav1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_342661.6 | XP_342662.5 | 3 | 1590 |