siRNA Details
Gene Symbol: Ndufb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108624.2 | NP_001102094.2 | 4 | 395 | ||
GenBank mRNA | BC166901.1 | - | 4 | 372 | 6922162 | |
BG668360.1 | - | 3 | 255 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ndufb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108624.2 | NP_001102094.2 | 4 | 395 | ||
GenBank mRNA | BC166901.1 | - | 4 | 372 | 6922162 | |
BG668360.1 | - | 3 | 255 |