siRNA Details
| Gene Symbol: Ndufb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108624.2 | NP_001102094.2 | 2, 3 | 286 | ||
| GenBank mRNA | BC166901.1 | - | 2, 3 | 263 | 6922162 | |
| BG668360.1 | - | 1, 2 | 146 | |||
| C06706.1 | - | 2, 3 | 287 | |||

Ndufb2
Genomic Map