siRNA Details
| Gene Symbol: Tcaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749739.3 | XP_003749787.1 | 3 | 987 | ||
| XM_008762928.1 | XP_008761150.1 | 3 | 1316 | |||
| XM_342675.6 | XP_342676.4 | 3 | 1316 | |||
| GenBank mRNA | FQ211472.1 | - | 3 | 954 | ||

Tcaf1
Genomic Map