siRNA Details
| Gene Symbol: Cul1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108627.2 | NP_001102097.1 | 22 | 2517 | ||
| XM_006236411.1 | XP_006236473.1 | 21 | 2425 | |||
| GenBank mRNA | BC161932.1 | - | 22 | 2517 | 9021933 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cul1| Gene Symbol: Cul1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108627.2 | NP_001102097.1 | 22 | 2517 | ||
| XM_006236411.1 | XP_006236473.1 | 21 | 2425 | |||
| GenBank mRNA | BC161932.1 | - | 22 | 2517 | 9021933 | |