siRNA Details
| Gene Symbol: Cul1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108627.2 | NP_001102097.1 | 14 | 1775 | ||
| XM_006236409.1 | XP_006236471.1 | 13 | 1683 | |||
| XM_006236410.2 | XP_006236472.1 | 13 | 1685 | |||
| XM_006236411.1 | XP_006236473.1 | 13 | 1683 | |||
| GenBank mRNA | BC161932.1 | - | 14 | 1775 | 9021933 | |

Cul1
Genomic Map