siRNA Details
| Gene Symbol: Hnrnpa2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104613.1 | NP_001098083.1 | 9 | 932 | ||
| GenBank mRNA | AB006815.1 | - | 8 | 896 | ||
| AB006816.1 | - | 9 | 932 | |||
| AB006817.1 | - | 7 | 776 | |||
| AB006818.1 | - | 8 | 812 | |||
| BC166548.1 | - | 407 | ||||

Hnrnpa2b1
Genomic Map