siRNA Details
| Gene Symbol: ASTN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286164.1 | NP_001273093.1 | 13 | 2432 | ||
| NM_004319.2 | NP_004310.1 | 13 | 2432 | |||
| NM_207108.2 | NP_996991.1 | 13 | 2432 | |||
| XM_017001340.1 | XP_016856829.1 | 13 | 2406 | |||
| XM_017001341.1 | XP_016856830.1 | 13 | 2406 | |||
| GenBank mRNA | AB006627.1 | - | 13 | 2230 | ||
| AK295126.1 | - | 13 | 2384 | |||
| AK309094.1 | - | 13 | 2501 | |||
| BC015438.1 | - | 1 | 76 | |||
| BC142697.1 | - | 13 | 2211 | 40148746 | ||
| BC146795.1 | - | 13 | 2230 | 9007266 | ||

ASTN1
Genomic Map