siRNA Details
| Gene Symbol: Lancl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014187.1 | NP_001014209.1 | 1, 2 | 221 | ||
| GenBank mRNA | BC079412.1 | - | 1, 2 | 221 | 7113320 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lancl2| Gene Symbol: Lancl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014187.1 | NP_001014209.1 | 1, 2 | 221 | ||
| GenBank mRNA | BC079412.1 | - | 1, 2 | 221 | 7113320 | |