siRNA Details
| Gene Symbol: Herc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108631.1 | NP_001102101.1 | 16 | 2421 | ||
| XM_006236584.2 | XP_006236646.1 | 15 | 2317 | |||
| XM_006236585.2 | XP_006236647.1 | 15 | 2317 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Herc3| Gene Symbol: Herc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108631.1 | NP_001102101.1 | 16 | 2421 | ||
| XM_006236584.2 | XP_006236646.1 | 15 | 2317 | |||
| XM_006236585.2 | XP_006236647.1 | 15 | 2317 | |||