siRNA Details
| Gene Symbol: Fam13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100862.1 | NP_001094332.1 | 13 | 1601 | ||
| XM_006236586.2 | XP_006236648.1 | 12 | 1608 | |||
| XM_008762964.1 | XP_008761186.1 | 13 | 1635 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam13a| Gene Symbol: Fam13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100862.1 | NP_001094332.1 | 13 | 1601 | ||
| XM_006236586.2 | XP_006236648.1 | 12 | 1608 | |||
| XM_008762964.1 | XP_008761186.1 | 13 | 1635 | |||