siRNA Details
| Gene Symbol: Rpia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108632.1 | NP_001102102.1 | 8 | 801 | ||
| XM_006236628.2 | XP_006236690.1 | 7 | 739 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpia| Gene Symbol: Rpia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108632.1 | NP_001102102.1 | 8 | 801 | ||
| XM_006236628.2 | XP_006236690.1 | 7 | 739 | |||