siRNA Details
Gene Symbol: Pole4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108634.1 | NP_001102104.1 | 4 | 913 | ||
GenBank mRNA | BC166842.1 | - | 2 | 1716 | 9028705 | |
FQ213769.1 | - | 4 | 887 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pole4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108634.1 | NP_001102104.1 | 4 | 913 | ||
GenBank mRNA | BC166842.1 | - | 2 | 1716 | 9028705 | |
FQ213769.1 | - | 4 | 887 |