siRNA Details
| Gene Symbol: LOC103692169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763084.1 | XP_008761306.1 | 1 | 317 | ||
| Gene Symbol: Mrpl53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108635.2 | NP_001102105.1 | 1 | 47 | ||
| GenBank mRNA | BC166928.1 | - | 1 | 47 | 6920305 | |

LOC103692169
Genomic Map