siRNA Details
| Gene Symbol: Tmem43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007745.1 | NP_001007746.1 | 12 | 1166 | ||
| GenBank mRNA | BC083626.1 | - | 12 | 1166 | 7134239 | |
| FQ221746.1 | - | 13 | 1327 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem43| Gene Symbol: Tmem43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007745.1 | NP_001007746.1 | 12 | 1166 | ||
| GenBank mRNA | BC083626.1 | - | 12 | 1166 | 7134239 | |
| FQ221746.1 | - | 13 | 1327 | |||