siRNA Details
| Gene Symbol: Sumf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108639.1 | NP_001102109.1 | 2, 3 | 442 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sumf1| Gene Symbol: Sumf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108639.1 | NP_001102109.1 | 2, 3 | 442 | ||