siRNA Details
| Gene Symbol: Zfp637 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134908.1 | NP_001128380.1 | 3 | 266 | ||
| XM_006237192.2 | XP_006237254.1 | 4 | 953 | |||
| XM_006237193.2 | XP_006237255.1 | 4 | 982 | |||
| XM_006237194.2 | XP_006237256.1 | 4 | 908 | |||
| GenBank mRNA | BC158822.1 | - | 3 | 266 | 7376045 | |
| FQ213084.1 | - | 3 | 238 | |||

Zfp637
Genomic Map