siRNA Details
Gene Symbol: Mfap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108644.1 | NP_001102114.1 | 10 | 511 | ||
GenBank mRNA | FQ229611.1 | - | 9 | 617 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mfap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108644.1 | NP_001102114.1 | 10 | 511 | ||
GenBank mRNA | FQ229611.1 | - | 9 | 617 |