siRNA Details
| Gene Symbol: RGD1307916 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108645.1 | NP_001102115.1 | 3 | 307 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1307916| Gene Symbol: RGD1307916 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108645.1 | NP_001102115.1 | 3 | 307 | ||