siRNA Details
| Gene Symbol: Lpcat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012189.1 | NP_001012189.1 | 8 | 891 | ||
| GenBank mRNA | BC089869.1 | - | 8 | 891 | 7324068 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lpcat3| Gene Symbol: Lpcat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012189.1 | NP_001012189.1 | 8 | 891 | ||
| GenBank mRNA | BC089869.1 | - | 8 | 891 | 7324068 | |