siRNA Details
| Gene Symbol: Gpr162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108646.1 | NP_001102116.1 | 2 | 1114 | ||
| XM_006237371.2 | XP_006237433.1 | 2 | 1211 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr162| Gene Symbol: Gpr162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108646.1 | NP_001102116.1 | 2 | 1114 | ||
| XM_006237371.2 | XP_006237433.1 | 2 | 1211 | |||