siRNA Details
| Gene Symbol: Iffo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108647.1 | NP_001102117.1 | 7 | 1417 | ||
| XM_006237372.2 | XP_006237434.1 | 8 | 1457 | |||
| XM_006237373.2 | XP_006237435.1 | 8 | 1454 | |||
| XM_006237374.2 | XP_006237436.1 | 8 | 1455 | |||
| XM_006237375.2 | XP_006237437.1 | 7 | 1448 | |||
| XM_006237376.2 | XP_006237438.1 | 7 | 1445 | |||
| XM_008763292.1 | XP_008761514.1 | 8 | 1482 | |||

Iffo1
Genomic Map