siRNA Details
| Gene Symbol: Ndufa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100752.1 | NP_001094222.1 | 5 | 473 | ||
| XM_006237500.1 | XP_006237562.1 | 5 | 556 | |||
| GenBank mRNA | BC091192.1 | - | 5 | 439 | ||
| BC168777.1 | - | 5 | 473 | 6922152 | ||
| FQ216233.1 | - | 5 | 473 | |||
| FQ216540.1 | - | 5 | 473 | |||
| FQ216671.1 | - | 5 | 463 | |||
| FQ216764.1 | - | 5 | 463 | |||
| FQ232722.1 | - | 5 | 471 | |||

Ndufa9
Genomic Map