siRNA Details
| Gene Symbol: Itfg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108648.1 | NP_001102118.1 | 8 | 915 | ||
| XM_006237409.2 | XP_006237471.1 | 8 | 1304 | |||
| GenBank mRNA | BC158714.1 | - | 7 | 770 | 9088606 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itfg2| Gene Symbol: Itfg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108648.1 | NP_001102118.1 | 8 | 915 | ||
| XM_006237409.2 | XP_006237471.1 | 8 | 1304 | |||
| GenBank mRNA | BC158714.1 | - | 7 | 770 | 9088606 | |