siRNA Details
| Gene Symbol: Klrb1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010964.1 | NP_001010964.1 | 3 | 248 | ||
| XM_006237414.2 | XP_006237476.1 | 3 | 308 | |||
| GenBank mRNA | EF100677.1 | - | 3 | 232 | ||
| EF100681.1 | - | 3 | 232 | |||
| EF100683.1 | - | 3 | 232 | |||
| FJ416340.1 | - | 3 | 253 | |||
| FQ232553.1 | - | 2 | 111 | |||
| M62891.1 | - | 3 | 248 | |||
| Gene Symbol: RGD2301395 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135687.1 | NP_001129159.1 | 3 | 232 | ||
| GenBank mRNA | FJ158033.1 | - | 3 | 232 | ||

Klrb1a
Genomic Map