siRNA Details
| Gene Symbol: Crebl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015027.1 | NP_001015027.1 | 3 | 460 | ||
| GenBank mRNA | BC091323.1 | - | 3 | 460 | 7313598 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crebl2| Gene Symbol: Crebl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015027.1 | NP_001015027.1 | 3 | 460 | ||
| GenBank mRNA | BC091323.1 | - | 3 | 460 | 7313598 | |