siRNA Details
| Gene Symbol: Hebp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108651.1 | NP_001102121.1 | 3 | 413 | ||
| XM_006237529.1 | XP_006237591.1 | 3 | 422 | |||
| GenBank mRNA | BC168221.1 | - | 3 | 371 | 7323058 | |
| FQ220508.1 | - | 3 | 413 | |||
| FQ229464.1 | - | 3 | 413 | |||
| FQ233110.1 | - | 3 | 382 | |||
| FQ235087.1 | - | 3 | 382 | |||

Hebp1
Genomic Map