siRNA Details
| Gene Symbol: Arhgdib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009600.1 | NP_001009600.1 | 3, 4 | 329 | ||
| XM_006237561.2 | XP_006237623.1 | 3, 4 | 353 | |||
| XM_006237562.2 | XP_006237624.1 | 3, 4 | 484 | |||
| XM_006237563.2 | XP_006237625.1 | 4, 5 | 415 | |||
| XM_006237564.2 | XP_006237626.1 | 3, 4 | 444 | |||
| XM_006237565.2 | XP_006237627.1 | 3, 4 | 629 | |||
| GenBank mRNA | BC088209.1 | - | 3, 4 | 329 | 7382227 | |
| FQ225418.1 | - | 3, 4 | 303 | |||
| FQ233135.1 | - | 3, 4 | 310 | |||
| FQ234136.1 | - | 3, 4 | 324 | |||

Arhgdib
Genomic Map