siRNA Details
| Gene Symbol: Fgfr1op2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201421.1 | NP_958824.1 | 4 | 688 | ||
| XM_006237688.2 | XP_006237750.1 | 4 | 677 | |||
| XM_008763402.1 | XP_008761624.1 | 4 | 667 | |||
| GenBank mRNA | AY426739.1 | - | 4 | 688 | ||
| AY426740.1 | - | 4 | 688 | |||
| BC087696.1 | - | 4 | 585 | 7312267 | ||

Fgfr1op2
Genomic Map