siRNA Details
| Gene Symbol: Arntl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF327071.1 | - | 15 | 1657 | ||
| AY014837.1 | - | 14 | 1555 | |||
| AY014838.1 | - | 14 | 1564 | |||
| JQ361086.1 | - | 15 | 1678 | |||
| Gene Symbol: LOC102555341 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763468.1 | XP_008761690.1 | 1222 | |||

Arntl2
Genomic Map