siRNA Details
| Gene Symbol: Cspp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006237767.2 | XP_006237829.1 | 17, 18 | 2203 | ||
| XM_006237778.2 | XP_006237840.1 | 17, 18 | 2200 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cspp1| Gene Symbol: Cspp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006237767.2 | XP_006237829.1 | 17, 18 | 2203 | ||
| XM_006237778.2 | XP_006237840.1 | 17, 18 | 2200 | |||