siRNA Details
| Gene Symbol: Tcea1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025735.1 | NP_001020906.1 | 2 | 162 | ||
| XM_006237816.2 | XP_006237878.1 | 2 | 223 | |||
| XM_008763546.1 | XP_008761768.1 | 2 | 50 | |||
| GenBank mRNA | BC099141.1 | - | 2 | 162 | 7460791 | |
| FQ227501.1 | - | 2 | 134 | |||
| Gene Symbol: LOC498453 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006251720.2 | XP_006251782.1 | 1 | 216 | ||

Tcea1
Genomic Map