siRNA Details
| Gene Symbol: MGC94199 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007746.1 | NP_001007747.1 | 531 | |||
| XM_006237871.2 | XP_006237933.1 | 876 | ||||
| GenBank mRNA | BC079168.1 | - | 531 | 7129828 | ||
| Gene Symbol: LOC102554232 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006237865.2 | XP_006237927.1 | 5 | 876 | ||
| XM_006237866.2 | XP_006237928.1 | 5 | 873 | |||

MGC94199
Genomic Map