siRNA Details
| Gene Symbol: Ccne2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108656.1 | NP_001102126.1 | 10 | 1044 | ||
| XM_006237873.1 | XP_006237935.1 | 10 | 1588 | |||
| XM_006237874.1 | XP_006237936.1 | 10 | 1051 | |||
| XM_006237875.1 | XP_006237937.1 | 9 | 919 | |||

Ccne2
Genomic Map