siRNA Details
| Gene Symbol: Ripk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191865.1 | NP_001178794.1 | 6 | 934 | ||
| GenBank mRNA | BC107468.1 | - | 4 | 365 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ripk2| Gene Symbol: Ripk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191865.1 | NP_001178794.1 | 6 | 934 | ||
| GenBank mRNA | BC107468.1 | - | 4 | 365 | ||