siRNA Details
| Gene Symbol: Dctn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108659.1 | NP_001102129.1 | 2, 3 | 211 | ||
| GenBank mRNA | FQ222216.1 | - | 2, 3 | 193 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dctn3| Gene Symbol: Dctn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108659.1 | NP_001102129.1 | 2, 3 | 211 | ||
| GenBank mRNA | FQ222216.1 | - | 2, 3 | 193 | ||