siRNA Details
| Gene Symbol: Anks6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015028.2 | NP_001015028.2 | 12 | 2333 | ||
| XM_008763699.1 | XP_008761921.1 | 12 | 2204 | |||
| XM_008763700.1 | XP_008761922.1 | 12 | 2204 | |||
| GenBank mRNA | AY661303.1 | - | 12 | 2333 | ||

Anks6
Genomic Map