siRNA Details
Gene Symbol: Msantd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108664.1 | NP_001102134.1 | 3 | 643 | ||
XM_008763701.1 | XP_008761923.1 | 2 | 884 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Msantd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108664.1 | NP_001102134.1 | 3 | 643 | ||
XM_008763701.1 | XP_008761923.1 | 2 | 884 |