siRNA Details
| Gene Symbol: Smc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108666.3 | NP_001102136.2 | 21, 22 | 3104 | ||
| XM_006238165.2 | XP_006238227.1 | 20, 21 | 3048 | |||
| GenBank mRNA | CV074555.1 | - | 1, 2 | 21 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smc2| Gene Symbol: Smc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108666.3 | NP_001102136.2 | 21, 22 | 3104 | ||
| XM_006238165.2 | XP_006238227.1 | 20, 21 | 3048 | |||
| GenBank mRNA | CV074555.1 | - | 1, 2 | 21 | ||