siRNA Details
| Gene Symbol: Zfp462 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006238178.2 | XP_006238240.1 | 3 | 4047 | ||
| XM_008763726.1 | XP_008761948.1 | 2 | 8764 | |||
| XM_008763727.1 | XP_008761949.1 | 3 | 6752 | |||
| XM_008763728.1 | XP_008761950.1 | 2 | 8764 | |||
| XM_008763729.1 | XP_008761951.1 | 2 | 8764 | |||

Zfp462
Genomic Map