siRNA Details
| Gene Symbol: Fkbp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763840.1 | XP_008762062.1 | 12 | 1409 | ||
| GenBank mRNA | FQ230831.1 | - | 11 | 1330 | ||
| Gene Symbol: LOC102556352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006238232.2 | XP_006238294.1 | 5 | 1025 | ||

Fkbp15
Genomic Map