siRNA Details
| Gene Symbol: Akna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108668.1 | NP_001102138.1 | 3 | 1373 | ||
| XM_006238255.2 | XP_006238317.1 | 2 | 1239 | |||
| XM_006238256.2 | XP_006238318.1 | 3 | 1718 | |||
| XM_006238257.2 | XP_006238319.1 | 3 | 1284 | |||
| XM_008763780.1 | XP_008762002.1 | 3 | 1286 | |||
| XM_008763781.1 | XP_008762003.1 | 4 | 3706 | |||
| XM_008763782.1 | XP_008762004.1 | 4 | 3167 | |||
| XM_008763783.1 | XP_008762005.1 | 3 | 2427 | |||
| XM_008763784.1 | XP_008762006.1 | 3 | 3036 | |||

Akna
Genomic Map