siRNA Details
| Gene Symbol: RGD1560884 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763844.1 | XP_008762066.1 | 11, 12 | 1636 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1560884| Gene Symbol: RGD1560884 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763844.1 | XP_008762066.1 | 11, 12 | 1636 | ||